Anti-GABA Guinea Pig Polyclonal Antibody

Supplier: Enzo Life Sciences
GZ1081-0025
89160-008EA 373.23 USD
89160-008
Anti-GABA Guinea Pig Polyclonal Antibody
Antibodies
Host: Guinea pig

  • For Immunohistochemistry

Recognizes GABA Species independent.

Type: Primary
Antigen: GABA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Guinea Pig
Isotype:
Reactivity: Species Independent
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR