ITSA-1 ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: (1H-Benzo[d][1,2,3]triazol-1-yl)(2,4-dichlorophenyl)methanone

BML-GR350-0025
89158-988EA 115.96 USD
89158-988
ITSA-1 ≥98% (by TLC)
ITSA-1

HDAC inhibitor


ITSA-1 is a cell permeable benzotriazole amide that counteracts trichostatin A (Prod. No. BML-GR309) induced cell cycle arrest, histone acetylation and transcriptional activation but not the activity of other HDAC inhibitors.

Formula: C₁₃H₇Cl₂N₃O
MW: 292.1202 g/mol
MDL Number: MFCD00195103
CAS Number: 200626-61-5
https://us.vwr.com/stibo/web/std.lang.all/12/70/45401270.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR