Amifostine ≥98% (by HPLC)
Supplier: Enzo Life Sciences
|
Warning
|
BML-GR341-0050
BML-GR341-0010
89158-976EA
419.28
USD
89158-976
89158-974
Amifostine ≥98% (by HPLC)
Amifostine
HIF-1α activator.
Amifostine is a broad-spectrum radio- and chemoprotector. It restores transcriptional activity of specific p53 mutants, prevents chemotherapy-induced apoptosis and upregulates HIF-1 & alpha.
Soluble in water.
Formula:
C₅H₁₅N₂O₃PS Storage Temperature: Refrigerator |
CAS Number:
20537-88-6 |
Specification Test Results
Purity | ≥98% (HPLC) |
Identity | Determined by IR |
Appearance | White to off-white crystalline powder |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...