Tomatidine HCl ≥90% (by TLC)
Supplier: Enzo Life Sciences
BML-GR335-0025
89158-960EA
84.38
USD
89158-960
Tomatidine HCl ≥90% (by TLC)
Tomatidine HCl
Negative control for cyclopamine.
Steroidal alkaloid structurally similar to cyclopamine but does not inhibit the hedgehog (shh) pathway. Used as a negative control for jervine.
Soluble in DMSO (2 mg/mL), 100% ethanol (2.5 mg/mL) or dimethyl formamide (2 mg/mL).
Formula:
C₂₇H₄₅NO₂·HCl |
CAS Number:
6192-62-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...