SBHA ≥95% (by TLC)
Supplier: Enzo Life Sciences
Synonyms:
Suberoyl bis-hydroxamic acid
BML-GR323-0500
BML-GR323-0100
89158-922EA
150.09
USD
89158-922
89158-920
SBHA ≥95% (by TLC)
SBHA
Induces cellular differentiation. Inhibits histone deacetylation.
- HDAC inhibitor
White to off-white solid.
Soluble in DMSO.
Formula:
C₈H₁₆N₂O₄ MW: 204.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00192455 CAS Number: 38937-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...