Mitomycin C ≥95%
Supplier: Enzo Life Sciences
Synonyms:
Ametycin
, Mitomycin
BML-GR311-0010
BML-GR311-0002
89158-880EA
208.31
USD
89158-880
89158-878
Mitomycin C ≥95%
Mitomycin C
Antitumor antibiotic
- Blue-violet solid
DNA crosslinking and damaging agent. Inhibitor of DNA synthesis, nuclear division and cancer cells. DNA intercalating agent. Induces apoptosis in a variety of cells via a p53-dependent pathway.
Soluble in water (1 mg/mL, warm), 100% ethanol (1 mg/mL, warm), DMSO (15 mg/mL) or methanol
Formula:
C₁₅H₁₈N₄O₅ MW: 334.33 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00078109 CAS Number: 50-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...