β-Lapachone ≥98%

Supplier: Enzo Life Sciences

Synonyms: 2,2-Dimethyl-3,4-dihydro-2H-benzo[h]chromene-5,6-dione

BML-GR308-0025 BML-GR308-0005
89158-870EA 108.24 USD
89158-870 89158-872
β-Lapachone ≥98%
beta-Lapachone

Topoisomerase inhibitor.


Novel DNA topoisomerase I inhibitor. Unlike camptothecin, beta-lapachone does not stabilize the cleavable complex, indicating a novel mode of action. Induces apoptosis in HL-60 and human prostate cancer cells via a p53-independent mechanism and blocks the cell cycle at G0/G1.

Formula: C₁₅H₁₄O₃
MW: 242.274 g/mol
MDL Number: MFCD01712233
CAS Number: 4707-32-8
https://us.vwr.com/stibo/web/std.lang.all/39/46/45403946.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR