Mithramycin A (from Streptomyces argillaceus) ≥95% (by HPLC, TLC)

Supplier: Enzo Life Sciences

Synonyms: Plicamycin

BML-GR305-0001
89158-864EA 81.24 USD
89158-864
Mithramycin A (from Streptomyces argillaceus) ≥95% (by HPLC, TLC)
Mithramycin A

Mithramycin noncovalently interacts with G-C-rich duplex DNA in the presence of Mg2+ or Zn2+


It induces differentiation of leukemic cells accompanied by an early decrease in c-myc expression. It selectively inhibits collagen-1 gene expression in human fibroblasts.


Yellow solid


Soluble in phosphate buffered saline (1 mg/mL), 100% ethanol, DMSO or methanol


Packaging: Shipping temperature: Ambient

Formula: C₅₂H₇₆O₂₄
Storage Temperature: Freezer
MDL Number: MFCD00135618
CAS Number: 18378-89-7
https://us.vwr.com/stibo/web/std.lang.all/39/44/45403944.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR