3-Methylcholanthrene ≥97% (by TLC)
BML-GR239-0010
BML-GR239-0050
89158-832EA
367.41
USD
89158-832
89158-830
3-Methylcholanthrene ≥97% (by TLC)
3-Methylcholanthrene
Aryl hydrocarbon receptor (AhR) agonist. Inhibits osteoclast formation by inhibiting the expression of receptor activator of NF-κB receptor ligand (RANKL). Suppresses growth factor and JAK-2 expression in mice.
- Ah receptor ligand
Yellow solid.
Soluble in chloroform or ethyl acetate.
Formula:
C₂₁H₁₆ MW: 268.4 g/mol Storage Temperature: Ambient |
CAS Number:
56-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...