Z-Guggulsterone ≥98%
Supplier: Enzo Life Sciences
BML-GR235-0005
89158-822EA
179.39
USD
89158-822
Z-Guggulsterone ≥98%
Z-Guggulsterone
A guggul tree steroid well known for its lipid lowering properties. Acts as a selective antagonist at farnesoid X receptors (FXRs). Inhibits FXR transactivation (IC50=10µM).
- FXR inhibitor
White solid.
Soluble in DMSO.
Formula:
C₂₁H₂₈O₂ MW: 312.5 g/mol Storage Temperature: Freezer |
CAS Number:
39025-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...