24(S)-Hydroxycholesterol ≥98% (by TLC), white solid
Supplier: Enzo Life Sciences
BML-GR230-0001
BML-GR230-0005
89158-808EA
618.67
USD
89158-808
89158-806
24(S)-Hydroxycholesterol ≥98% (by TLC), white solid
24(S)-Hydroxycholesterol
LXR ligand.
Endogenous agonist for the nuclear receptor LXR. Important metabolite of cholesterol in the brain. Displays neurotoxic effects on human neuroblastoma cells. Cerebrospinal fluid concentrations of 24(S)-hydroxycholesterol are increased in patients with Alzheimer’s disease and mild cognitive impairment.
Soluble in ethanol (10 mg/mL) or DMSO (10 mg/mL).
Formula:
C₂₇H₄₆O₂ Melting Pt: 177…180 °C Storage Temperature: Freezer |
CAS Number:
474-73-7 |
Specification Test Results
Purity | ≥98% (TLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...