Troglitazone ≥98% (by TLC)
Supplier: Enzo Life Sciences
Synonyms:
5-(4-((6-Hydroxy-2,5,7,8-tetramethylchroman-2-yl)methoxy)benzyl)thiazolidine-2,4-dione
PPAR ligand
Antidiabetic thiazolidinedione. Potent and selective PPARγ ligand. Binds to the PPARγ ligand-binding domain and recruits ligand-specific transcriptional coactivators at 1µM. In skeletal muscle and adipocyte cell cultures, the EC50 of troglitazone is about 10µM. Displaces [125I]SB-236636 with an IC50=7.9 µM. Stimulates adipogenesis and decreases osteoblastogenesis in human mesenchymal stem cells. Induces apoptosis in human vascular smooth muscle cells and inhibits the growth of a variety of human cancer cells including lung, bladder and myeloid leukemia cells.
Synthetic.
Formula:
C₂₄H₂₇NO₅S MW: 441.5 g/mol |
MDL Number:
MFCD00878416 CAS Number: 97322-87-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...