6-Formylindolo[3,2-b]carbazole ≥95% (by HPLC), yellow solid
Supplier: Enzo Life Sciences
|
Warning
|
Synonyms:
5,11-Dihydroindolo[3,2-b]carbazole-6-carbaldehyde
BML-GR206-0100
89158-788EA
138.95
USD
89158-788
6-Formylindolo[3,2-b]carbazole ≥95% (by HPLC), yellow solid
6-Formylindolo[3,2-b]carbazole
Ah receptor ligand.
A tryptophan-derived, high affinity aryl hydrocarbon receptor ligand (Kd=7×10-11 M). Proposed to be an endogenous AhR ligand. Induces rapid and transient expression of cytochrome P450-1A1 (CYP1A1) in human cells at 100 pM.
Soluble in DMSO (10 mg/mL).
Formula:
C₁₉H₁₂N₂O MW: 284.32 g/mol Melting Pt: >300 °C Storage Temperature: Freezer |
MDL Number:
MFCD23140892 CAS Number: 172922-91-7 |
Specification Test Results
Purity | ≥95% (HPLC) |
Appearance | Yellow solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...