Methoprene ≥98% (by TLC)
Supplier: Enzo Life Sciences
|
Warning
|
BML-GR107-0010
89158-760EA
101.43
USD
89158-760
Methoprene ≥98% (by TLC)
Methoprene
Retinoic acid receptor ligand.
Methoprene is an analog of the insect growth regulator, juvenile hormone. Retinoic acid receptor ligand after ester hydrolysis.
Soluble in DMSO or ethanol.
Formula:
C₁₉H₃₄O₃ |
CAS Number:
40596-69-8 UN: 3082 ADR: 9,III |
Specification Test Results
Purity | ≥98% (TLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...