Methoprene acid ≥98% (by TLC)
Supplier: Enzo Life Sciences
BML-GR106-0005
89158-756EA
133.71
USD
89158-756
Methoprene acid ≥98% (by TLC)
Methoprene acid
Retinoid X receptor ligand.
Methoprene acid is an analog of the insect growth regulator, juvenile hormone. It binds directly to RXR and is a transcriptional activator in both insect and mammalian cells. It is RXR specific and does not activate the RAR pathway.
Soluble in DMSO or methanol.
Formula:
C₁₆H₂₈O₃ MW: 268.397 g/mol |
MDL Number:
MFCD01317817 CAS Number: 53092-52-7 |
Specification Test Results
Purity | ≥98% (TLC) |
Appearance | ale yellow oil |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...