Blocking Peptide For Glutamic Acid Decarboxylase mAb (11) (Prod. No. BmL-GC3208) Blocking Peptide
Supplier: Enzo Life Sciences
GP3209-0050
89158-728EA 137.73 USD
89158-728
Blocking Peptide For Glutamic Acid Decarboxylase mAb (11) (Prod. No. BmL-GC3208) Blocking Peptide
Proteins and Peptides
Immunization and blocking peptide for Glutamic Acid Decarboxylase (GAD) antibody (BML-GC3208). The tetradecapeptide corresponds to the N-terminal residues 4-17 of rat GAD65. The sequence is wholly conserved human GAD65.