Blocking Peptide For Glutamic Acid Decarboxylase pAb (Prod. No. BmL-GC3008) Blocking Peptide
Supplier: Enzo Life Sciences
GP3009-0050
89158-726EA 137.73 USD
89158-726
Blocking Peptide For Glutamic Acid Decarboxylase pAb (Prod. No. BmL-GC3008) Blocking Peptide
Proteins and Peptides
Immunization and blocking peptide for Glutamic Acid Decarboxylase (GAD) antibodies (BML-GC3008 and BML-GC3108). The pentadecapeptide corresponds to the C-terminal residues [Cys+]572-585 of rat GAD65. The sequence is wholly conserved at the C-terminus of rat GAD67, mouse, pig and human GAD65/67, cat GAD67, and in human GAD-2.