Would you like to log in again?
Glucagon like peptide 1 (human) (7-36) amide, Enzo Life Sciences
Supplier: Enzo Life Sciences
GP1438-0100
A predominant cleaved form of pro-glucagon expressed in the intestine. GLP-1 [7-36]amide is promptly released from the gut after oral ingestion of a meal where it slows gastric emptying and exerts a powerful insulinotropic action leading to elevated serum insulin levels. GLP-1 [7-36] amide promotes satiety and reduces food intake in patients with diabetes mellitus type 2 and has been suggested to have therapeutic potential in the treatment of NIDDM. It has been reported that human pancreatic endocrine tumors contain molecular forms of GLP-1 which resemble those expressed in the small intestine, rather than in their tissue of origin. Serves as a control peptide for Anti-GLP-1 [7-36] Amide (BML-GA1178). Blocks immunoreactivity at 10 nmol per ml diluted antibody.
ORDER
Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg GP1438-0100 89158-722 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Protein/Peptide Name Glucagon like peptide 1 (7-36) amide
Species Human
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty