Isotetrandrine ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-G520-0001
89158-648EA 90.18 USD
89158-648
Isotetrandrine ≥98% (by TLC)
Isotetrandrine

Phospholipase inhibitor


Isotetrandrine is a biscoclaurine alkaloid which inhibits G-protein mediated activation of phospholipase A2 (PLA2) but not phospholipase C (PLC) or D (PLD). Cell permeable.

Formula: C₃₈H₄₂N₂O₆
MW: 622.762 g/mol
MDL Number: MFCD00236404
CAS Number: 477-57-6
https://us.vwr.com/stibo/web/std.lang.all/38/93/45403893.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR