Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ilimaquinone ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-G435-0100
89158-638
Ilimaquinone ≥98% (by TLC)
ADP-ribosylation factor inhibitor

Displays antimicrobial, anti-HIV and anti-inflammatory properties. Induces complete and reversible breakdown of Golgi membranes. Inhibits the association of β-COP and ADP-ribosylation factor to Golgi membranes, but unlike BFA, does not induce retrograde transport of Golgi enzymes into ER or cause random fusion of successive Golgi cisternae. Interacts with methylation enzymes. Shows anticancer activity.


Formula: C₂₂H₃₀O₄
MW: 358.478 g/mol
CAS Number: 71678-03-0
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty