u-83836e ≥98% (by HPLC), white to off-white solid

Supplier: Enzo Life Sciences

BML-FR101-0100
89158-472EA 158.43 USD
89158-472
u-83836e ≥98% (by HPLC), white to off-white solid
u-83836e

Antioxidant.


The lazaroid prevents hydrogen peroxide-induced F₂ isoprostane production in renal proximal tubular cells and reduces secondary brain injury in a rabbit model of cryogenic trauma.


Soluble in water (30 mg/mL) or ethanol (>50 mg/mL).

Formula: C₃₀H₄₄N₆O₂
MW: 520.722 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09750018
CAS Number: 122003-11-6
https://us.vwr.com/stibo/web/std.lang.all/38/59/45403859.jpg

Order Now

Specification Test Results

Purity ≥98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR