Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FN1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
FG6010-0100
Fibronectins are high molecular weight, disulphide-linked, dimeric cell adhesion glycoproteins found in basement membranes and in the interstitial connective tissue matrix. A single fibronectin gene is subject to alternative splicing in a cell-type-, development- and age-regulated manner which gives rise to multiple molecular forms. In addition to their prominent role in adhesion, fibronectins have been reported to mediate various aspects of cellular interaction, including migration during development and wound-healing, haemostasis, and the regulation of cell growth and differentiation. Cellular fibronectins (cFn) are found in low amounts in normal human plasma and tissues, but they are abundant in the plasma of carcinoma patients and in the stroma of various carcinomas. In contrast, a soluble form of fibronectin produced by hepatocytes is readily detectable in plasma and becomes deposited in pericellular matrices and within tissues. This form of fibronectin, referred to as ‘plasma fibronectin’ (pFn), differs from cFn by the absence of an amino acid sequence, known as extra domain A1.

For Immunocytochemistry, Western Blot

Recognizes the extra domain A (ED) of cellular fibronectin (cFn). Detects a band of ~ 250kDa by Western blot. Recognizes cFN in malignant tumors, does not cross-react with benign tumors or normal tissues.

Type: Primary
Antigen: FIBRONECTIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL FG6010-0100 89158-458 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol FN1
Antigen Name Fibronectin
Reactivity Human
Clonality Monoclonal
Antibody Type Primary
Host Mouse
Isotype IgG1
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms ED-B, GFND2, MSF, GFND, FN, Cold insoluble globulin, LETS, CIG, FINC, FNZ, fibronectin 1
Purification Protein A affinity chromatography
Form Liquid. In PBS containing 0.1% sodium azide and 1mg/mL BSA.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty