Docosa-7Z,10Z,13Z,16Z,19Z-pentaenoic acid ≥98%

Supplier: Enzo Life Sciences

Synonyms: Clupanodonic acid(7Z,10Z,13Z,16Z,19Z)-docosa-7,10,13,16,19-pentaenoic acid

BML-FA012-0010
89158-346EA 88.09 USD
89158-346
Docosa-7Z,10Z,13Z,16Z,19Z-pentaenoic acid ≥98%
Docosa-7Z,10Z,13Z,16Z,19Z-pentaenoic acid

Polyunsaturated fatty acid.


Soluble in ethanol, hexane and methylene chloride.

Formula: C₂₂H₃₄O₂
MW: 330.512 g/mol
CAS Number: 24880-45-3
https://us.vwr.com/stibo/web/std.lang.all/38/41/45403841.jpg

Order Now

Specification Test Results

Purity ≥98%
Appearance Clear to pale yellow oil

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR