STA-21 ≥98% (by TLC)
Supplier: Enzo Life Sciences
BML-EI351-0001
89158-156EA 286.63 USD
89158-156
STA-21 ≥98% (by TLC)
STA-21
STAT inhibitor
STA-21 is a novel Stat3-selective inhibitor. It inhibits Stat3 dimerization and DNA binding as well as Stat3-dependent luciferase reporter activity.
CAS Number: 28882-53-3
https://us.vwr.com/stibo/web/std.lang.all/37/94/45403794.jpg