URB-602 ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: Biphenyl-3-yl carbamic acid cyclohexyl ester

BML-EI348-0050 BML-EI348-0010
89158-144EA 92.55 USD
89158-144 89158-146
URB-602 ≥98% (by TLC)
URB-602

Selectively inhibits monoacylglycerol (MAG) lipase (IC50=28µM) via a noncompetitive mechanism. Does not inhibit FAAH, DAG lipase or COX-2. URB-602 selectively raises levels of 2-arachidonoylglycerol with no change in anandamide concentrationsin cells and tissues. In the same way anandamide levels were markely elevated by the FAAH inhibitor URB597 at 1µM.


  • Monoacylglycerol lipase inhibitor


Off-white solid.


Soluble in DMSO (30 mg/ml) or ethanol (5 mg/ml warm).

Formula: C₁₉H₂₁NO₂
MW: 295.4 g/mol
Melting Pt: 122…123 °C
Storage Temperature: Ambient
CAS Number: 565460-15-3
https://us.vwr.com/stibo/web/std.lang.all/37/92/45403792.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR