BIBB-515 ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-EI340-0010 BML-EI340-0050
89158-120EA 525.88 USD
89158-120 89158-118
BIBB-515 ≥98% (by TLC)
BIBB-515

BIBB 515 is a potent and selective inhibitor of 2,3-oxidosqualene cyclase (OSC) in vivo. BIBB 515 inhibits LDL production in both normolipemic and hyperlipemic hamsters and may represent a novel approach for the development of lipid lowering drugs.


  • Oxido-squalene cyclase inhibitor


White solid.


Soluble in DMSO (2mg/ml) or DMF (8mg/ml).

Formula: C₂₂H₂₁ClN₂O₂
MW: 380.9 g/mol
Storage Temperature: Ambient
CAS Number: 156635-05-1
https://us.vwr.com/stibo/web/std.lang.all/37/85/45403785.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR