SB202190 ≥98% (by TLC)
Supplier: Enzo Life Sciences
Synonyms:
4-(4-(4-Fluorophenyl)-5-(pyridin-4-yl)-1H-imidazol-2-yl)phenol
BML-EI294-0001
89157-986EA
143.84
USD
89157-986
SB202190 ≥98% (by TLC)
SB202190
Cell permeable, potent and selective p38 MAP kinase inhibitor (IC50=30 nM). Inhibits p38α and ß, but not γ and δ isoforms. Does not inhibit ERK2 or other members of the MAP kinase family or their upstream activators. A useful tool for dissecting the role of p38 in signaling pathways. Induces apoptosis.
- p38 inhibitor
Pale yellow solid.
Soluble in methanol, DMSO or acetone; slightly soluble in ethyl acetate.
Formula:
C₂₀H₁₄FN₃O MW: 331.3 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00941964 CAS Number: 152121-30-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...