(S)-(+)-Ketoprofen ≥98.5% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms: (S)-2-(3-Benzoylphenyl)propanoic acid
BML-EI288-0001
89157-974EA 237.13 USD
89157-974
(S)-(+)-Ketoprofen ≥98.5% (by HPLC)
(S)-(+)-Ketoprofen
Active enantiomer of ketoprofen. Inhibits cyclooxygenase.
  • COX inhibitor
White solid.
Soluble in 100% ethanol (25mg/ml).
https://us.vwr.com/stibo/web/std.lang.all/37/40/45403740.jpg