Bisindolylmaleimide IX·methanesulfonate ≥98%

Supplier: Enzo Life Sciences

Synonyms: Ro 31-8220 mesylate

BML-EI283-0001
89157-964EA 127.69 USD
89157-964
Bisindolylmaleimide IX·methanesulfonate ≥98%
Bisindolylmaleimide IX·methanesulfonate

Cell-permeable PKC inhibitor. Selective for PKC (IC50=10nM) over CaM kinase II (IC50=17µM) and PKA (IC50=0.9µM). Induces apoptosis in various cell lines.


  • PKC inhibitor


Red/orange solid.


Soluble in DMSO (10mg/ml)

Formula: C₂₅H₂₃N₅O₂S·CH₄O₃S
MW: 457.6·96.1 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03788207
CAS Number: 138489-18-6
https://us.vwr.com/stibo/web/std.lang.all/37/37/45403737.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR