RG-13022 ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: 3-(3,4-Dimethoxyphenyl)-2-(pyridin-3-yl)acrylonitrileTyrphostin RG 13022

BML-EI252-0005
89157-874EA 129.52 USD
89157-874
RG-13022 ≥98% (by TLC)
RG-13022

RG-13022 is a non-phenolic tyrphostin analog, which inhibits the EGF receptor (IC50=5 µM) and PDGF receptor tyrosine kinases, and is long-acting. It inhibits EGF-stimulated HER14 cell proliferation (IC50=1 µM) as well as tumor growth in vivo. It inhibits both EGF- and PDGF-stimulated DNA synthesis in human glioma cell lines. RG-13022 is non-phenolic and therefore possesses less antioxidant activity than other tyrphostins.


  • Tyrosine kinase inhibitor


White solid.


Soluble at 25mg/ml in DMSO or 100% ethanol.

Formula: C₁₆H₁₄N₂O₂
MW: 266.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236444
CAS Number: 136831-48-6
https://us.vwr.com/stibo/web/std.lang.all/37/13/45403713.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR