Benzylphosphonic acid-(AM)2 ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-EI245-0010
89157-856EA 94.35 USD
89157-856
Benzylphosphonic acid-(AM)2 ≥98% (by TLC)
Benzylphosphonic acid-(AM)2

Benzylphosphonic acid-(AM)2 is a bis-acetoxymethyl ester (AM) derivative of benzylphosphonic acid which is cell permeable and is converted to the active phosphonic acid via intracellular esterase-catalyzed cleavage of the AM esters.


  • PTP inhibitor


White solid.


Soluble in ethanol (20 mg/ml) or DMSO (20 mg/ml).

Formula: C₁₃H₁₇O₇P
MW: 316.3 g/mol
Melting Pt: 38…39 °C
Storage Temperature: Freezer
CAS Number: 200716-68-3
https://us.vwr.com/stibo/web/std.lang.all/37/08/45403708.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR