K-252b ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-EI234-1000 BML-EI234-0100
89157-840EA 713.97 USD
89157-840 89157-838
K-252b ≥98% (by HPLC)
K-252b

General, cell permeable protein kinase inhibitor. Acts by binding to the ATP binding domain of the kinase. Potent inhibitor of Ca2+/calmodulin kinase II. Inhibits myosin light chain kinase (MLCK; Ki=0.147µM), cAMP- dependent protein kinase (PKA; Ki=0.09µM), protein kinase C (PKC; Ki=0.02µM), and cGMP-dependent protein kinase (PKG; Ki=0.1µM).


  • PKC inhibitor


White solid.


Soluble in DMSO (1mg/ml) or dimethyl formamide (1mg/ml).

Formula: C₂₆H₁₉N₃O₅
MW: 453.5 g/mol
Storage Temperature: Refrigerator
CAS Number: 99570-78-2
https://us.vwr.com/stibo/web/std.lang.all/37/05/45403705.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR