AG-494 ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: Tyrphostin AG 494(E)-2-Cyano-3-(3,4-dihydroxyphenyl)-N-phenylacrylamide

BML-EI228-0010
89157-816EA 98.54 USD
89157-816
AG-494 ≥98% (by TLC)
AG-494

AG-494 is a member of the tyrphostin family of tyrosine kinase inhibitors and is a potent inhibitor of EGF receptor autophosphorylation (IC50=1.2 µM) and EGF-dependent cell growth (IC50=6 µM). It selectively inhibits HER1 (EGF receptor) vs. HER1-2 receptor autophosphorylation (HER1: IC50=1.1 µM; HER1-2: IC50=45 µM). HER1-2 is a chimeric receptor consisting of the external HER1 domain fused to an internal HER2 domain. Blocks CDK2 activation and causescells to arrest at late G1 and during S phase.


  • EGFR inhibitor


Yellow solid.


Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml).

Formula: C₁₆H₁₂N₂O₃
MW: 280.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00209863
CAS Number: 133550-35-3
https://us.vwr.com/stibo/web/std.lang.all/36/99/45403699.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR