KT5823 ≥98% (by TLC)
Supplier: Enzo Life Sciences
BML-EI200-0100
89157-772EA 190.48 USD
89157-772
KT5823 ≥98% (by TLC)
KT5823
PKG inhibitor
KT5823 is a derivative of K-252a and is a selective, cell permeable, inhibitor of PKG (Ki=0.234 µM). Ki’s for PKA =4μM and for MLCK=>10μM.
CAS Number: 126643-37-6
https://us.vwr.com/stibo/web/std.lang.all/36/90/45403690.jpg