KT5720 ≥98% (by TLC)
Supplier: Enzo Life Sciences
BML-EI199-0100
89157-770EA 200.93 USD
89157-770
KT5720 ≥98% (by TLC)
KT5720
PKA inhibitor
KT5720 is a hexylester derivative of K-252a and is a selective inhibitor of PKA (Ki=0.056 µM). Cell permeable.
CAS Number: 108068-98-0
https://us.vwr.com/stibo/web/std.lang.all/36/89/45403689.jpg