H-8 dihydrochloride ≥99% (by TLC)

Supplier: Enzo Life Sciences

BML-EI158-0050 BML-EI158-0010
89157-680EA 120.15 USD
89157-680 89157-682
H-8 dihydrochloride ≥99% (by TLC)
H-8 dihydrochloride

PKA inhibitor


H-8 inhibits PKA and PKG selectively over PKC and MLCK (IC50’s=1.2μM, 0.48μM, 15μM and 68μM respectively). It also inhibits myosin light chain kinase.

Formula: C₁₂H₁₅N₃O₂S·2HCl
MW: 265.3·72.9 g/mol
CAS Number: 113276-94-1
https://us.vwr.com/stibo/web/std.lang.all/36/65/45403665.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR