Oleyloxyethyl phosphorylcholine ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-EI118-0010
89157-600EA 94.35 USD
89157-600
Oleyloxyethyl phosphorylcholine ≥98% (by HPLC)
Oleyloxyethyl phosphorylcholine

A potent and specific substrate analog PLA2 inhibitor (IC50=6.2 µM-porcine pancreatic PLA2). It inhibits human recombinant sPLA2 (IC50=13.7 µM) selectively over cPLA2. At 1 µM it completely blocked the myogenic response in perfused dog renal arcuate arteries.


  • Phospholipase inhibitor


Colorless oil or waxy solid.


Soluble in 100% ethanol.

Formula: C₂₅H₅₂NO₅P
MW: 477.7 g/mol
Storage Temperature: Freezer
CAS Number: 96720-06-8
https://us.vwr.com/stibo/web/std.lang.all/36/26/45403626.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR