Oxatomide ≥99% (by HPLC)

Supplier: Enzo Life Sciences

BML-EI114-0010
89157-588EA 150.09 USD
89157-588
Oxatomide ≥99% (by HPLC)
Oxatomide

Suppresses PAF-induced bronchoconstriction in the guinea pig (ED50=0.9 mg/kg orally).


  • Inhibits the release and actions of leukotrienes


White solid.


Soluble in ethanol or DMSO.

Formula: C₂₇H₃₀N₄O
MW: 426.6 g/mol
Storage Temperature: Ambient
CAS Number: 60607-34-3
https://us.vwr.com/stibo/web/std.lang.all/36/22/45403622.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR