Leukotriene B4-dimethylamide ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-EI102-0050
89157-554EA 454.45 USD
89157-554
Leukotriene B4-dimethylamide ≥98% (by HPLC)
Leukotriene B4-dimethylamide

Leukotriene B4 antagonist. Inhibits binding of [3H]-LTB4 to guinea pig lung membranes (Ki=0.13 µM). Antagonizes the degranulation response to LTB4 in rabbit PMNL (Kd=0.2 µM) acting, most likely, via receptor desensitization. Partial agonist at higher concentrations (5 µM).


  • Prostanoid receptor ligand


Liquid.

Formula: C₂₂H₃₇NO₃
MW: 363.5 g/mol
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
CAS Number: 83024-92-4
https://us.vwr.com/stibo/web/std.lang.all/36/13/45403613.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR