BMOLEE14
BML-EE14-0050 BML-EE14-1000
89157-526EA 102.74 USD
89157-526 89157-528
BMOLEE14
COX inhibitor
Stimulates glucagon release from rat pancreatic islets. Inhibits cyclooxygenase and platelet aggregation. Inhibits vasopressin response. Induces monocyte adhesion to endothelial cells. Inhibits renin release. Decreases blood pressure in rats.