Ibotenic acid ≥98% (by HPLC), white to off-white solid

Supplier: Enzo Life Sciences

Synonyms: α-Amino-3-hydroxy-5-isoxazoleacetic acid2-Amino-2-(3-oxo-2,3-dihydroisoxazol-5-yl)acetic acid

BML-EA120-0005 BML-EA120-0001
89157-450EA 154.37 USD
89157-450 89157-452
Ibotenic acid ≥98% (by HPLC), white to off-white solid
Ibotenic acid

mGluR agonist. Potent glutamate receptor agonist. Displays full agonist activity at mGlu1 and mGlu5.


Soluble in water (20 mg/mL).


Caution: Store, as supplied, at room temperature for up to 1 year. Store solutions at -20 °C for up to 3 months.

Formula: C₅H₆N₂O₄
MW: 158.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00069294
CAS Number: 2552-55-8
UN: 3462
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/36/04/45403604.jpg

Order Now

Specification Test Results

Identity Determined by H-NMR and MS
Purity ≥98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR