5(S),6(R)-Dihydroxyeicosa-7E,9E,11Z,14Z-tetraenoic acid (DiHETE) ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 5(S),6(R)-DiHETE

BML-DS056-0050
89157-390EA 289.77 USD
89157-390
5(S),6(R)-Dihydroxyeicosa-7E,9E,11Z,14Z-tetraenoic acid (DiHETE) ≥99% (by HPLC)
5(S),6(R)-Dihydroxyeicosa-7E,9E,11Z,14Z-tetraenoic acid (DiHETE)

Synthesis via enzymatic and nonenzymatic hydrolysis of LTA4. Induces contraction of guinea pig lung parenchymal strips. Partial agonist at LTD4 receptors. Potent inhibitor of LTB4 induced neutrophil migration (IC50 = 8nM).


  • Prostanoid receptor ligand


Colorless liquid.

Formula: C₂₀H₃₂O₄
MW: 336.5 g/mol
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
CAS Number: 82948-88-7
https://us.vwr.com/stibo/web/std.lang.all/35/98/45403598.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR