5(S),15(S)-DIHETE ≥99% (by HPLC)
Supplier: Enzo Life Sciences
|
Danger
|
BML-D515-0050
89157-314EA
222.53
USD
89157-314
5(S),15(S)-DIHETE ≥99% (by HPLC)
5(S),15(S)-DIHETE
Arachidonic acid metabolite.
Biosynthesis in human eosinophils. Enhances neutrophil degranulation. Potent chemotaxin for human eosinophils.
Formula:
C₂₀H₃₂O₄ Storage Temperature: Ultra-Low Temperature Freezer (–80°C) |
CAS Number:
82200-87-1 EINECS: 200-578-6 UN: 1170 ADR: 3,II |
Specification Test Results
Concentration | 50 µg/mL |
Formulation | Oil dissolved in ethanol |
Purity | ≥99% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...