BTCP ≥98% (by HPLC)

BML-D103-0010
89157-280EA 278.59 USD
89157-280
BTCP ≥98% (by HPLC)
BTCP

Potent dopamine re-uptake inhibitor.


  • Dopamine receptor inhibitor


Off-white solid.


Soluble in 100% ethanol.

Formula: C₁₉H₂₅NS·C₄H₄O₄
MW: 415.6 g/mol
Storage Temperature: Refrigerator
CAS Number: 112726-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR