LY2183240 ≥98% (by TLC)
Supplier: Enzo Life Sciences
BML-CR113-0050 BML-CR113-0010
89157-136EA 404.69 USD
89157-136 89157-134
LY2183240 ≥98% (by TLC)
LY2183240
Anandamide transporter inhibitor
A potent inhibitor of anandamide uptake (IC50=7.3nM). Systemic administration to rodents elevates brain anandamide levels and shows efficacy in a rodent model of persistent pain. LY2183240 also inhibits FAAH hydrolytic activity (IC50=12.4nM) as well as other brain serine proteases and represents a new chemotype for the design of new selective FAAH inhibitors. Radiolabeled LY2183240 has been used to identify a high-affinity, saturable anandamide transporter binding site on RBL-2H3 cell membranes distinct from FAAH.
CAS Number: 874902-19-9
https://us.vwr.com/stibo/web/std.lang.all/35/73/45403573.jpg