MDL-12,330A ≥98% (by HPLC)

Synonyms: cis-N-(2-Phenylcyclopentyl)azacyclotridec-1-en-2-amine hydrochloride

BML-CN145-0005
89157-022EA 84.94 USD
89157-022
MDL-12,330A ≥98% (by HPLC)
MDL-12,330A

Adenylate cyclase inhibitor (IC50=250 µM).


  • Adenosine receptor inhibitor


White solid.


Soluble in water (3mg/ml), DMSO (10mg/ml, slightly warm), 100% ethanol (>10mg/ml) and hot 0.1N HCl (0.5mg/ml); insoluble in 0.1N NaOH.

Formula: C₂₃H₃₆N₂·HCl
MW: 377 g/mol
Storage Temperature: Freezer
CAS Number: 40297-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR