PK 11195 ≥98% (by TLC)
Supplier: Enzo Life Sciences
BML-CM118-0005
BML-CM118-0025
89156-956EA
347.91
USD
89156-956
89156-954
PK 11195 ≥98% (by TLC)
PK 11195
Benzodiazepine receptor ligand.
PK-11195 is a ligand of the mitochondrial benzodiazepine receptor and selective antagonist of benzodiazepine. It facilitates the induction of apoptosis and reverses Bcl-2-mediated cytoprotection.
Soluble in 100% ethanol, methanol, dichlormethane or DMSO. Insoluble in water.
Formula:
C₂₁H₂₁ClN₂O |
CAS Number:
85532-75-8 |
Specification Test Results
Purity | ≥98% (HPLC) |
Identity | Determined by ¹H-NMR and MS |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...