BMOLCC100
CC100-0500 BML-CC100-0100
89156-894EA 362.85 USD
89156-894 89156-892
BMOLCC100
p58 PITSLREbeta1 inhibitor
A purine analog that can override multiple cell cycle checkpoints. May be used to override drug-induced cell cycle blocks. It is a selective protein kinase inhibitor that can inhibit p58 PITSLREbeta1, a p34cdc2-related protein kinase, in vivo.