Gabapentin ≥99% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: Neurontin2-[1-(Aminomethyl)cyclohexyl]acetic acid

BML-CA234-0020 BML-CA234-0100
89156-806EA 406.78 USD
89156-806 89156-804
Gabapentin ≥99% (by TLC)
Gabapentin

Ca2+ channel blocker


Gabapentin is a GABA analog which does not act at GABA receptors but which inhibits L-type calcium channels via binding to the alpha2-delta subunit and neuropathic pain relieving actions. Ligands acting at the alpha2-delta subunit lead to anticonvulsant, anxiolytic and neuropathic pain-relieving actions.

Formula: C₉H₁₇NO₂
MW: 171.24 g/mol
MDL Number: MFCD00865286
CAS Number: 60142-96-3
https://us.vwr.com/stibo/web/std.lang.all/34/78/45403478.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR