Desipramine hydrochloride ≥98% (by TLC)
BML-AR119-1000 BML-AR119-0100
89156-638EA 62.53 USD
89156-638 89156-640
Desipramine hydrochloride ≥98% (by TLC)
Desipramine hydrochloride
Norepinephrine transporter inhibitor
A selective inhibitor of norepinephrine transporter (Ki=4nM). Serotonin and dopamine transporters are also inhibited at higher concentrations (Ki=61nM and 78.7µM respectively). Reverses the pro-apoptotic effect of chronic unpredictable mild stress in the rat cerebral cortex. Induces apoptosis in several cancer cell lines. Displays anti-inflammatory activity in a rat colitis model.
CAS Number: 58-28-6